Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXXC4
Entrez Gene ID
102121275
Gene Name
CXXC finger protein 4
Gene Aliases
-
Chromosome Location
Chr.5: 103896570 - 103920228 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005555592.2 XP_005555649.1 1 - 2 651 93

View Full Product Details