Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPO
Entrez Gene ID
102138508
Gene Name
carboxypeptidase O
Gene Aliases
-
Chromosome Location
Chr.12: 96538804 - 96574915 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005574078.2 XP_005574135.1 3 - 4 2989 88

View Full Product Details