Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102145718
Entrez Gene ID
102145718
Gene Name
cytochrome c oxidase assembly factor 7
Gene Aliases
SELRC1
Chromosome Location
Chr.1: 174794220 - 174801320 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005543353.2 XP_005543410.1 1 - 2 460 87

View Full Product Details