Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A1BG
Entrez Gene ID
102129055
Gene Name
alpha-1-B glycoprotein
Gene Aliases
-
Chromosome Location
Chr.19: 58937416 - 58951837 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015441069.1 XP_015296555.1 5 - 6 1026 63

View Full Product Details