Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
13000
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
1110035J23Rik, C77789, CK2
Chromosome Location
Chr.8: 95446096 - 95488904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009974.3 NP_034104.1 2 - 3 589 108
GenBank mRNA AF012251.1 - 2 - 3 314 108
AJ001420.1 - 2 - 3 559 108
AK002927.1 - 2 - 3 597 108
AK076042.1 - 2 - 3 550 108
BC057862.1 - 2 - 3 249 108

View Full Product Details