Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr1d2
Entrez Gene ID
353187
Gene Name
nuclear receptor subfamily 1, group D, member 2
Gene Aliases
RVR, Rev-erb
Chromosome Location
Chr.14: 18204056 - 18239106 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011584.4 NP_035714.3 1 - 2 295 60
GenBank mRNA AK054522.1 - 1 - 2 220 60
AK079075.1 - 1 - 2 182 60
BC096461.1 - 1 - 2 135 60
CJ079681.1 - 1 - 2 295 60
U12142.1 - 1 - 2 261 60

View Full Product Details