The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011905.3 | NP_036035.3 | 2 - 3 | 340 | 69 |
GenBank mRNA | AF124741.1 | - | 2 - 3 | 279 | 69 |
AF165189.1 | - | 2 - 3 | 304 | 69 | |
AF185284.1 | - | 2 - 3 | 294 | 69 | |
AF216289.1 | - | 2 - 3 | 303 | 69 | |
AK005043.1 | - | 2 - 3 | 341 | 69 | |
AK154504.1 | - | 2 - 3 | 310 | 69 | |
AY179346.1 | - | 1 - 2 | 155 | 69 | |
BC014693.1 | - | 2 - 3 | 499 | 69 | |
BY743020.1 | - | 2 - 3 | 308 | 69 |