Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map3k14
Entrez Gene ID
53859
Gene Name
mitogen-activated protein kinase kinase kinase 14
Gene Aliases
Nik, aly
Chromosome Location
Chr.11: 103219764 - 103267401 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016896.3 NP_058592.1 10 - 11 1911 64
GenBank mRNA AF143094.1 - 9 - 10 1828 64
AK036200.1 - 10 - 11 1912 64
AK172518.1 - 10 - 11 1881 64

View Full Product Details