Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cope
Entrez Gene ID
59042
Gene Name
coatomer protein complex, subunit epsilon
Gene Aliases
1110005D17Rik, Cope1
Chromosome Location
Chr.8: 70302785 - 70312990 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021538.1 NP_067513.1 4 - 5 468 88
GenBank mRNA AB039837.1 - 4 - 5 468 88
AK003458.1 - 4 - 5 485 88
AK140753.1 - 2 - 3 2189 88
BC009170.1 - 4 - 5 476 88
BC057328.1 - 2 - 3 2154 88
BC083336.1 - 4 - 5 450 88

View Full Product Details