The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024226.4 | NP_077188.1 | 3 - 4 | 669 | 85 |
NM_194051.3 | NP_918940.1 | 5 - 6 | 2956 | 85 | |
NM_194052.3 | NP_918941.1 | 3 - 4 | 1145 | 85 | |
NM_194053.3 | NP_918942.1 | 4 - 5 | 1202 | 85 | |
NM_194054.3 | NP_918943.1 | 5 - 6 | 3563 | 85 | |
GenBank mRNA | AB073672.1 | - | 3 - 4 | 1521 | 85 |
AF326337.1 | - | 3 - 4 | 449 | 85 | |
AK220511.1 | - | 5 - 6 | 3523 | 85 | |
AY102280.1 | - | 5 - 6 | 2956 | 85 | |
AY102281.1 | - | 3 - 4 | 1102 | 85 | |
AY102282.1 | - | 4 - 5 | 1159 | 85 | |
AY102283.1 | - | 3 - 4 | 662 | 85 | |
AY102284.1 | - | 5 - 6 | 3520 | 85 | |
AY114152.1 | - | 5 - 6 | 3521 | 85 | |
AY114153.1 | - | 4 - 5 | 976 | 85 | |
BC032192.1 | - | 3 - 4 | 1815 | 85 | |
BC032272.1 | - | 3 - 4 | 2669 | 85 | |
BC056373.1 | - | 5 - 6 | 3391 | 85 | |
BU848611.1 | - | 3 - 4 | 508 | 85 |