The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015781.4 | NP_056596.1 | 2 - 3 | 190 | 125 |
GenBank mRNA | AK004633.1 | - | 2 - 3 | 160 | 125 |
AK007322.1 | - | 2 - 3 | 155 | 125 | |
AK008261.1 | - | 1 - 2 | 53 | 125 | |
AK032896.1 | - | 2 - 3 | 163 | 125 | |
AK136161.1 | - | 2 - 3 | 156 | 125 | |
AK145766.1 | - | 2 - 3 | 154 | 125 | |
AK152011.1 | - | 2 - 3 | 83 | 125 | |
AK153531.1 | - | 2 - 3 | 83 | 125 | |
AK169301.1 | - | 2 - 3 | 102 | 125 | |
D12618.1 | - | 2 - 3 | 146 | 125 |