Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pkp3
Entrez Gene ID
56460
Gene Name
plakophilin 3
Gene Aliases
2310056L12Rik
Chromosome Location
Chr.7: 141078229 - 141090510 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162924.1 NP_001156396.1 4 - 5 1114 67
NM_019762.2 NP_062736.2 3 - 4 1045 67
GenBank mRNA AF136719.1 - 3 - 4 1036 67
BC020472.1 - 3 - 4 1017 67
BC032045.1 - 3 - 4 1005 67
BC090668.1 - 4 - 5 1114 67
BC106141.1 - 3 - 4 1010 67
BC139775.1 - 3 - 4 1001 67

View Full Product Details