Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubxn4
Entrez Gene ID
67812
Gene Name
UBX domain protein 4
Gene Aliases
1300013G12Rik, Ubxd2, mKIAA0242
Chromosome Location
Chr.1: 128243898 - 128279378 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026390.2 NP_080666.2 10 - 11 1340 74
GenBank mRNA AK032206.1 - 10 - 11 1218 74
AK075716.1 - 10 - 11 1075 74
AK129098.1 - 1 - 2 114 74
AK145551.1 - 7 - 8 766 74
BC019795.1 - 10 - 11 1359 74

View Full Product Details