Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd5
Entrez Gene ID
66998
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 5
Gene Aliases
1500032A03Rik, AW475925, S5b, W91691, mKIAA0072
Chromosome Location
Chr.2: 34852089 - 34870962 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080554.2 NP_542121.2 9 - 10 1295 96
GenBank mRNA AK011026.1 - 9 - 10 1272 96
AK029067.1 - 7 - 8 2672 96
AK032992.1 - 9 - 10 1273 96
AK078419.1 - 9 - 10 1273 96
AK145587.1 - 9 - 10 1272 96
AK148284.1 - - 1296 96
AK208538.1 - 1 - 2 119 96
AK213478.1 - 1 - 2 119 96
AK220321.1 - 9 - 10 1236 96
BC010650.1 - 3 - 4 396 96
BC019112.1 - 9 - 10 1306 96

View Full Product Details