Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pigt
Entrez Gene ID
78928
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class T
Gene Aliases
2510012P17Rik, 4930534E15Rik, CGI-06, NDAP, Ndap7
Chromosome Location
Chr.2: 164497511 - 164508301 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133779.2 NP_598540.2 10 - 11 1450 61
GenBank mRNA AK019717.2 - 10 - 11 1440 61
AK044499.1 - 10 - 11 1434 61
AK049171.1 - 10 - 11 1444 61
AK153462.1 - 10 - 11 1436 61
AK153464.1 - 10 - 11 1436 61
AK155057.1 - 10 - 11 1437 61
AK157226.1 - 10 - 11 1430 61
BC016520.1 - 11 - 12 1504 61
BC034175.1 - 10 - 11 1426 61

View Full Product Details