The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199349.1 | NP_001186278.1 | 9 - 10 | 912 | 65 |
NM_138309.3 | NP_612182.1 | 8 - 9 | 843 | 65 | |
GenBank mRNA | AK031264.1 | - | 9 - 10 | 669 | 65 |
AK080225.1 | - | 9 - 10 | 668 | 65 | |
AK166275.1 | - | 9 - 10 | 663 | 65 | |
AY078163.1 | - | 8 - 9 | 639 | 65 | |
BC031736.1 | - | 8 - 9 | 618 | 65 | |
EF516990.1 | - | 9 - 10 | 670 | 65 |