The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282093.1 | NP_001269022.1 | 7 - 8 | 1139 | 78 |
NM_153085.4 | NP_694725.3 | 7 - 8 | 1298 | 78 | |
GenBank mRNA | AF320996.1 | - | 7 - 8 | 1078 | 78 |
AK087826.1 | - | 7 - 8 | 1055 | 78 | |
AK156780.1 | - | 7 - 8 | 1123 | 78 | |
AK172416.1 | - | 7 - 8 | 1122 | 78 | |
BC057178.1 | - | 7 - 8 | 931 | 77 | |
BC080851.1 | - | 7 - 8 | 1256 | 78 | |
CN668018.1 | - | 3 - 4 | 499 | 78 |