Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Edc3
Entrez Gene ID
353190
Gene Name
enhancer of mRNA decapping 3 homolog (S. cerevisiae)
Gene Aliases
AA517853, Lsm16, Yjdc
Chromosome Location
Chr.9: 57708534 - 57750167 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153799.3 NP_722494.1 5 - 6 1124 59
GenBank mRNA AK090005.1 - - 815 59
AK140127.1 - 4 - 5 819 59
AK146804.1 - 5 - 6 1154 59
AK166286.1 - 5 - 6 1121 59
BC031725.1 - 5 - 6 1124 59
BC033484.1 - 5 - 6 1125 59

View Full Product Details