Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpl39l
Entrez Gene ID
68172
Gene Name
ribosomal protein L39-like
Gene Aliases
4930517K11Rik
Chromosome Location
Chr.16: 10170192 - 10174911 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026594.2 NP_080870.1 1 - 2 100 81
GenBank mRNA AK005645.1 - 1 - 2 90 81
AK019681.1 - 1 - 2 102 81
BC023885.1 - 1 - 2 90 81

View Full Product Details