The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122993.1 | NP_001116465.1 | 4 - 4 | 845 | 79 |
NM_033149.3 | NP_149161.1 | 3 - 3 | 478 | 79 | |
GenBank mRNA | BC047347.1 | - | 4 - 4 | 845 | 79 |
BC051669.1 | - | 4 - 4 | 845 | 79 | |
BC057887.1 | - | 3 - 3 | 478 | 79 |