The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029836.2 | NP_001025007.1 | 1 - 2 | 325 | 62 |
NM_001287101.1 | NP_001274030.1 | 1 - 2 | 325 | 62 | |
NM_001287102.1 | NP_001274031.1 | 1 - 2 | 325 | 62 | |
NM_033525.3 | NP_277060.2 | 1 - 2 | 325 | 62 | |
GenBank mRNA | AB059656.1 | - | 1 - 2 | 325 | 62 |
AF397007.1 | - | 1 - 2 | 77 | 62 | |
AF397008.1 | - | 1 - 2 | 77 | 62 | |
AI527763.1 | - | 1 - 2 | 205 | 62 | |
AK050484.1 | - | 1 - 2 | 229 | 62 | |
AY035898.1 | - | 1 - 2 | 200 | 62 | |
AY035899.1 | - | 1 - 2 | 200 | 62 | |
CK387038.1 | - | 1 - 2 | 199 | 62 |