The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286607.1 | NP_001273536.1 | 6 - 7 | 1034 | 85 |
NM_053242.4 | NP_444472.2 | 8 - 9 | 1434 | 85 | |
NM_212435.1 | NP_997600.1 | 6 - 7 | 1097 | 85 | |
GenBank mRNA | AF339106.1 | - | 5 - 6 | 775 | 85 |
AK051668.1 | - | 8 - 9 | 1417 | 85 | |
AK082361.1 | - | 8 - 9 | 1106 | 85 | |
AY079003.1 | - | 8 - 9 | 1063 | 85 | |
BC058960.1 | - | 6 - 7 | 1031 | 85 | |
BC062926.1 | - | 6 - 7 | 1097 | 85 |