The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011166.2 | NP_035296.1 | 1 - 2 | 73 | 73 |
GenBank mRNA | AF011384.1 | - | 1 - 2 | 72 | 73 |
AF015563.1 | - | 1 - 2 | 73 | 73 |