Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plod3
Entrez Gene ID
26433
Gene Name
procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3
Gene Aliases
AI414586, LH3
Chromosome Location
Chr.5: 136987019 - 136996646 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011962.3 NP_036092.1 4 - 5 1179 99
GenBank mRNA AF046783.1 - 4 - 5 587 99
AK013195.1 - 4 - 5 725 99
AK033360.1 - 4 - 5 724 99
AK053320.1 - 4 - 5 821 99
AK088948.1 - 4 - 5 596 99
BC043047.1 - 4 - 5 1178 99
BC054734.1 - 4 - 5 1179 99

View Full Product Details