The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011962.3 | NP_036092.1 | 18 - 19 | 2743 | 62 |
GenBank mRNA | AF046783.1 | - | 18 - 19 | 2151 | 62 |
AK013195.1 | - | 18 - 19 | 2289 | 62 | |
AK033360.1 | - | 18 - 19 | 2288 | 62 | |
AK088948.1 | - | 18 - 19 | 2160 | 62 | |
BC043047.1 | - | 18 - 19 | 2742 | 62 | |
BC054734.1 | - | 18 - 19 | 2743 | 62 |