Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcsk1
Entrez Gene ID
18548
Gene Name
proprotein convertase subtilisin/kexin type 1
Gene Aliases
Nec-1, Nec1, PC1, PC3, Phpp-1, SPC3
Chromosome Location
Chr.13: 75089415 - 75134861 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013628.2 NP_038656.1 7 - 8 955 73
GenBank mRNA BC108982.1 - 7 - 8 933 73
BC108983.1 - 7 - 8 933 73
M55668.1 - 2 - 3 241 73
M58589.1 - 7 - 8 955 73
M69196.1 - 7 - 8 1061 73
X57088.1 - 7 - 8 1067 73

View Full Product Details