Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr.12: 81860028 - 82000924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018814.3 NP_061284.2 35 - 36 7239 91
GenBank mRNA AF096286.1 - 29 - 30 5069 91
AK014387.1 - 9 - 10 1927 91
AK129221.1 - 30 - 31 5119 91
AK154122.1 - 16 - 17 2967 91
AK170320.1 - 16 - 17 2967 91
BC066849.1 - 30 - 31 4902 91
BC098370.1 - 30 - 31 4829 91

View Full Product Details