The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021318.3 | NP_067293.1 | 3 - 4 | 516 | 73 |
GenBank mRNA | AF083394.1 | - | 3 - 4 | 464 | 73 |
AK006414.1 | - | 3 - 4 | 505 | 73 | |
BC048473.1 | - | 3 - 4 | 516 | 73 |