Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Matn3
Entrez Gene ID
17182
Gene Name
matrilin 3
Gene Aliases
AV009181
Chromosome Location
Chr.12: 8947929 - 8972028 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010770.4 NP_034900.4 3 - 4 1016 71
GenBank mRNA AK048456.1 - 3 - 4 974 71
AK160954.1 - 3 - 4 993 71
BC071224.1 - 3 - 4 949 71
Y10521.1 - 3 - 4 981 71

View Full Product Details