The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177950.1 | NP_001171421.1 | 4 - 5 | 528 | 75 |
NM_001177951.1 | NP_001171422.1 | 4 - 5 | 528 | 75 | |
NM_001177952.1 | NP_001171423.1 | 4 - 5 | 522 | 75 | |
NM_001177953.1 | NP_001171424.1 | 4 - 5 | 528 | 75 | |
NM_001177954.1 | NP_001171425.1 | 4 - 5 | 528 | 75 | |
NM_011285.2 | NP_035415.1 | 4 - 5 | 528 | 75 | |
GenBank mRNA | AF044677.1 | - | 4 - 5 | 446 | 75 |
AJ238396.1 | - | 4 - 5 | 440 | 75 | |
AK017192.1 | - | 4 - 5 | 460 | 75 | |
AK029864.1 | - | 4 - 5 | 654 | 75 | |
AK038652.1 | - | 4 - 5 | 492 | 75 | |
AK081631.1 | - | 4 - 5 | 467 | 75 | |
AK136298.1 | - | 4 - 5 | 459 | 75 | |
AK138977.1 | - | 4 - 5 | 523 | 75 | |
BC012661.1 | - | 4 - 5 | 467 | 75 |