The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025358.2 | NP_001020529.2 | 11 - 12 | 1554 | 83 |
NM_001025359.2 | NP_001020530.2 | 11 - 12 | 1554 | 83 | |
NM_001025360.2 | NP_001020531.2 | 11 - 12 | 1573 | 83 | |
NM_001025361.2 | NP_001020532.2 | 11 - 12 | 1554 | 83 | |
NM_001025362.2 | NP_001020533.2 | 11 - 12 | 1554 | 83 | |
NM_001025363.2 | NP_001020534.2 | 11 - 12 | 1554 | 83 | |
NM_001081959.1 | NP_001075428.1 | 11 - 12 | 1573 | 83 | |
NM_008450.2 | NP_032476.2 | 11 - 12 | 1573 | 83 | |
GenBank mRNA | AF055665.1 | - | 11 - 12 | 1469 | 83 |
AK014256.1 | - | 3 - 4 | 232 | 83 | |
AK031309.1 | - | 11 - 12 | 1545 | 83 | |
AY753300.1 | - | 10 - 11 | 1378 | 83 | |
BC055744.1 | - | 11 - 12 | 1409 | 83 | |
CK032061.1 | - | 5 - 6 | 478 | 83 | |
CX734724.1 | - | 3 - 4 | 83 |