Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnfaip6
Entrez Gene ID
21930
Gene Name
tumor necrosis factor alpha induced protein 6
Gene Aliases
TSG-6, Tnfip6, Tsg6
Chromosome Location
Chr.2: 52038113 - 52056681 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009398.2 NP_033424.1 5 - 6 725 71
GenBank mRNA AK139985.1 - 5 - 6 735 71
BC021155.1 - 5 - 6 704 71
U83903.1 - 5 - 6 725 71

View Full Product Details