The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204071.1 | NP_001191000.1 | 8 - 9 | 758 | 115 |
NM_009420.2 | NP_033446.1 | 8 - 9 | 776 | 115 | |
GenBank mRNA | BC049615.1 | - | 8 - 9 | 728 | 115 |
M25533.1 | - | 8 - 9 | 728 | 115 |