The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010691.5 | NP_034821.2 | 1 - 2 | 340 | 116 |
GenBank mRNA | AB108497.1 | - | 1 - 2 | 340 | 116 |
AK144562.1 | - | 1 - 2 | 185 | 116 | |
BC119177.1 | - | 1 - 2 | 429 | 116 | |
BC120586.1 | - | 1 - 2 | 429 | 116 |