The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016808.2 | NP_058088.2 | 3 - 4 | 1010 | 78 |
NM_198091.2 | NP_932759.1 | 2 - 3 | 466 | 78 | |
NM_198092.2 | NP_932760.2 | 3 - 4 | 1107 | 78 | |
GenBank mRNA | AB041799.1 | - | 2 - 3 | 427 | 78 |
AF079565.1 | - | 2 - 3 | 68 | 78 | |
AK138799.1 | - | 4 - 5 | 1308 | 78 | |
AY255637.1 | - | 2 - 3 | 435 | 78 | |
AY255638.1 | - | 3 - 4 | 989 | 78 | |
AY255639.1 | - | 3 - 4 | 943 | 78 | |
AY255640.1 | - | 3 - 4 | 988 | 78 | |
BC017517.1 | - | 3 - 4 | 989 | 78 |