Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tub
Entrez Gene ID
22141
Gene Name
tubby candidate gene
Gene Aliases
rd5, tub
Chromosome Location
Chr.7: 109010880 - 109034459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021885.4 NP_068685.1 6 - 7 903 114
GenBank mRNA AK147700.1 - 6 - 7 958 114
AK165720.1 - 6 - 7 904 114
BC096554.1 - 6 - 7 884 114
U52433.1 - 6 - 7 891 114
U52824.1 - 7 - 8 1036 114
U54643.1 - 6 - 7 821 114

View Full Product Details