Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcvrn
Entrez Gene ID
19674
Gene Name
recoverin
Gene Aliases
CAR, S-modulin
Chromosome Location
Chr.11: 67695326 - 67703355 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009038.2 NP_033064.1 2 - 3 578 72
GenBank mRNA BC110564.2 - 2 - 3 571 72
CK617354.1 - 2 - 3 538 72
CK617615.1 - 1 - 2 72
X66196.1 - 2 - 3 586 72

View Full Product Details