Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc7a10
Entrez Gene ID
53896
Gene Name
solute carrier family 7 (cationic amino acid transporter, y+ system), member 10
Gene Aliases
AL024237, Asc-1, D7Bwg0847e
Chromosome Location
Chr.7: 35186357 - 35201114 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017394.4 NP_059090.3 8 - 9 1230 69
GenBank mRNA AB026688.1 - 8 - 9 1144 69
AK005282.1 - 6 - 7 627 69
AK031867.1 - 8 - 9 1198 69
BC054765.1 - 8 - 9 1173 69

View Full Product Details