Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Appl1
Entrez Gene ID
72993
Gene Name
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1
Gene Aliases
2900057D21Rik, 7330406P05Rik, AI585782, AW209077, BB022931, C88264, DIP13
Chromosome Location
Chr.14: 26918986 - 26971205 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145221.2 NP_660256.1 18 - 19 1790 62
GenBank mRNA AK030445.1 - 18 - 19 1669 62
AK033566.1 - 18 - 19 1761 62
AK045438.1 - 18 - 19 1726 62
AK049307.1 - 18 - 19 1777 62
AK146356.1 - 18 - 19 1761 62
AK173169.1 - 3 - 4 241 62
AY113705.1 - 18 - 19 1755 62
BC019708.1 - 10 - 11 1068 62
BC063751.1 - 18 - 19 1802 62

View Full Product Details