The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025832.2 | NP_080108.1 | 5 - 6 | 585 | 145 |
GenBank mRNA | AK005056.1 | - | 5 - 6 | 595 | 145 |
AK028432.1 | - | 5 - 6 | 696 | 146 | |
AK152498.1 | - | 5 - 6 | 595 | 145 | |
BC052445.1 | - | 5 - 6 | 585 | 145 |