Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Naa16
Entrez Gene ID
66897
Gene Name
N(alpha)-acetyltransferase 16, NatA auxiliary subunit
Gene Aliases
1300019C06Rik, Narg1l
Chromosome Location
Chr.14: 79334507 - 79390777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025832.2 NP_080108.1 7 - 8 860 144
GenBank mRNA AK005056.1 - 7 - 8 870 144
AK007755.1 - 7 - 8 896 144
AK028432.1 - 7 - 8 971 144
AK152498.1 - 7 - 8 870 144
BC052445.1 - 7 - 8 860 144

View Full Product Details