The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243584.1 | NP_001230513.1 | 1 - 2 | 186 | 89 |
NM_001243587.1 | NP_001230516.1 | 1 - 2 | 186 | 89 | |
GenBank mRNA | AK010175.1 | - | 1 - 2 | 140 | 89 |
AK150749.1 | - | 1 - 2 | 140 | 89 | |
BC019515.1 | - | 1 - 2 | 156 | 89 | |
BC026740.1 | - | 1 - 2 | 146 | 89 | |
BC026745.1 | - | 1 - 2 | 104 | 89 | |
CX210298.1 | - | 1 - 2 | 170 | 89 |