Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scara5
Entrez Gene ID
71145
Gene Name
scavenger receptor class A, member 5 (putative)
Gene Aliases
4932433F15Rik, 4933425F03Rik, AV278087, Tesr
Chromosome Location
Chr.14: 65666403 - 65764826 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168318.1 NP_001161790.1 2 - 3 463 76
NM_028903.2 NP_083179.2 2 - 3 463 76
GenBank mRNA AK016912.1 - 2 - 3 430 76
AK033154.1 - 2 - 3 430 76
AK133176.1 - 2 - 3 464 76
BC016096.1 - 2 - 3 439 76
BC023907.1 - 2 - 3 379 76
BC032159.1 - 2 - 3 419 76
DQ122126.1 - 1 - 2 113 76

View Full Product Details