The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082976.2 | NP_001076445.1 | 9 - 10 | 1216 | 81 |
NM_001286364.1 | NP_001273293.1 | 9 - 10 | 1879 | 81 | |
NM_028924.3 | NP_083200.1 | 9 - 10 | 1568 | 81 | |
GenBank mRNA | AB062282.1 | - | 8 - 9 | 1048 | 81 |
AK016683.1 | - | 9 - 10 | 1900 | 81 | |
AK136558.1 | - | 9 - 10 | 1217 | 81 | |
BC120793.1 | - | 9 - 10 | 1568 | 81 |