Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arfrp1
Entrez Gene ID
76688
Gene Name
ADP-ribosylation factor related protein 1
Gene Aliases
1500006I01Rik, AI480700
Chromosome Location
Chr.2: 181357690 - 181365404 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165991.1 NP_001159463.1 5 - 6 555 76
NM_001165992.1 NP_001159464.1 5 - 6 536 76
NM_001165995.1 NP_001159467.1 4 - 5 442 76
NM_029702.4 NP_083978.3 5 - 6 549 76
GenBank mRNA AJ413953.1 - 5 - 6 502 76
AK005174.1 - 5 - 6 497 76
AK044759.1 - 5 - 6 507 76
AK050136.1 - 2 - 3 1407 76
AK185500.1 - 2 - 3 155 76
BC010713.1 - 5 - 6 496 76

View Full Product Details