The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029956.3 | NP_084232.1 | 1 - 2 | 150 | 111 |
GenBank mRNA | AK020286.1 | - | 1 - 2 | 150 | 111 |
AK034288.1 | - | 1 - 2 | 115 | 111 | |
AK088785.1 | - | 1 - 2 | 142 | 111 | |
AK142286.1 | - | 1 - 2 | 142 | 111 | |
AK148421.1 | - | 1 - 2 | 185 | 111 | |
BC022159.1 | - | 1 - 2 | 104 | 111 | |
BC057558.1 | - | 1 - 2 | 150 | 111 |