Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crmp1
Entrez Gene ID
12933
Gene Name
collapsin response mediator protein 1
Gene Aliases
CRMP-1, DRP-1, Dpysl1, ULIP-3, Ulip3
Chromosome Location
Chr.5: 37242058 - 37292163 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136058.2 NP_001129530.1 10 - 11 1539 68
NM_007765.4 NP_031791.3 10 - 11 1404 68
GenBank mRNA AB006714.1 - 10 - 11 1376 68
AK159049.1 - 10 - 11 1348 68
BC031738.1 - 10 - 11 1333 68
BC065046.1 - 10 - 11 1517 68
U72875.1 - 10 - 11 1149 68
Y09080.1 - 10 - 11 1379 68

View Full Product Details