The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008102.3 | NP_032128.1 | 5 - 6 | 731 | 60 |
GenBank mRNA | AK150657.1 | - | 5 - 6 | 743 | 60 |
AK152571.1 | - | 5 - 6 | 743 | 60 | |
BC005643.1 | - | 5 - 6 | 731 | 60 | |
BC069921.1 | - | 5 - 6 | 735 | 60 | |
L09737.1 | - | 5 - 6 | 718 | 60 |