The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013483.3 | NP_038511.1 | 3 - 4 | 593 | 75 |
GenBank mRNA | AK142596.1 | - | 3 - 4 | 500 | 75 |
AK145168.1 | - | 3 - 4 | 594 | 75 | |
BC011497.1 | - | 3 - 4 | 567 | 75 | |
BC031459.1 | - | 2 - 3 | 491 | 75 | |
S80642.1 | - | 1 - 2 | 319 | 75 |