The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145432.3 | NP_663407.2 | 11 - 12 | 1814 | 65 |
GenBank mRNA | AK076219.1 | - | 11 - 12 | 1942 | 65 |
AK132494.1 | - | 11 - 12 | 1749 | 65 | |
AK149059.1 | - | 11 - 12 | 1609 | 65 | |
BC003942.1 | - | 4 - 5 | 529 | 65 | |
BC065092.1 | - | 11 - 12 | 1740 | 65 |